Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ADM
Entrez Gene ID
280713
Gene Name
adrenomedullin
Gene Aliases
-
Chromosome Location
Chr.15: 42377559 - 42379886 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173888.3 NP_776313.1 1 - 2 126 91
GenBank mRNA BC123826.1 - 1 - 2 126 91

View Full Product Details