Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GLRB
Entrez Gene ID
281198
Gene Name
glycine receptor beta
Gene Aliases
-
Chromosome Location
Chr.17: 42021083 - 42110395 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174071.2 NP_776496.1 2 - 3 194 116
GenBank mRNA AF268376.1 - 1 - 2 237 116
BC123491.1 - 2 - 3 194 116

View Full Product Details