Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TF
Entrez Gene ID
280705
Gene Name
transferrin
Gene Aliases
-
Chromosome Location
Chr.1: 135579069 - 135618070 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177484.3 NP_803450.2 10 - 11 1407 141
GenBank mRNA BC122602.1 - 10 - 11 1407 141
U02564.1 - 10 - 11 1354 141

View Full Product Details