Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPADH2
Entrez Gene ID
527722
Gene Name
spermadhesin 2
Gene Aliases
-
Chromosome Location
Chr.26: 42517620 - 42524490 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025338.2 NP_001020509.2 3 - 4 242 86
GenBank mRNA AJ853856.2 - 3 - 4 242 86
AJ853857.2 - 3 - 4 242 86

View Full Product Details