Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STAP1
Entrez Gene ID
507296
Gene Name
signal transducing adaptor family member 1
Gene Aliases
-
Chromosome Location
Chr.6: 83291634 - 83322915 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077868.1 NP_001071336.1 2 - 3 230 112
GenBank mRNA BC126666.1 - 2 - 3 230 112

View Full Product Details