Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM243
Entrez Gene ID
614280
Gene Name
transmembrane protein 243
Gene Aliases
C4H7orf23, C7orf23
Chromosome Location
Chr.4: 33227016 - 33248626 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078044.1 NP_001071512.1 4 - 5 599 74
GenBank mRNA BC126746.1 - 4 - 5 599 74

View Full Product Details