Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BMERB1
Entrez Gene ID
617444
Gene Name
bMERB domain containing 1
Gene Aliases
C25H16orf45
Chromosome Location
Chr.25: 13881226 - 14026630 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078077.1 NP_001071545.1 1 - 2 165 78
GenBank mRNA BC126524.1 - 1 - 2 165 78

View Full Product Details