Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KIF3C
Entrez Gene ID
777770
Gene Name
kinesin family member 3C
Gene Aliases
-
Chromosome Location
Chr.11: 73459329 - 73495510 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078155.1 NP_001071623.1 5 - 6 2371 67
GenBank mRNA BC126505.1 - 5 - 6 2371 67

View Full Product Details