Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C4H7orf57
Entrez Gene ID
614195
Gene Name
chromosome 4 C7orf57 homolog
Gene Aliases
C7orf57
Chromosome Location
Chr.4: 7668402 - 7697286 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079785.1 NP_001073253.1 3 - 4 409 98
GenBank mRNA BC126755.1 - 3 - 4 409 98

View Full Product Details