Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C18H19orf47
Entrez Gene ID
531855
Gene Name
chromosome 18 C19orf47 homolog
Gene Aliases
-
Chromosome Location
Chr.18: 49685146 - 49710176 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080291.1 NP_001073760.1 7 - 8 747 55
GenBank mRNA BC126749.1 - 7 - 8 747 55

View Full Product Details