Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC6A20
Entrez Gene ID
532774
Gene Name
solute carrier family 6 member 20
Gene Aliases
-
Chromosome Location
Chr.22: 53510186 - 53569209 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080294.2 NP_001073763.1 6 - 7 1003 123
GenBank mRNA BC126662.1 - 6 - 7 1004 123

View Full Product Details