Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NUDT4
Entrez Gene ID
614183
Gene Name
nudix hydrolase 4
Gene Aliases
-
Chromosome Location
Chr.5: 23191665 - 23208110 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081618.1 NP_001075087.1 3 - 4 592 84
GenBank mRNA BC133414.1 - 3 - 4 592 84

View Full Product Details