Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EFEMP1
Entrez Gene ID
511566
Gene Name
EGF containing fibulin extracellular matrix protein 1
Gene Aliases
-
Chromosome Location
Chr.11: 38484919 - 38554429 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081717.2 NP_001075186.1 7 - 8 989 69
GenBank mRNA BC133561.1 - 7 - 8 991 69

View Full Product Details