Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SIGIRR
Entrez Gene ID
531801
Gene Name
single Ig and TIR domain containing
Gene Aliases
-
Chromosome Location
Chr.29: 50869452 - 50877585 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082443.2 NP_001075912.1 8 - 9 1220 71
GenBank mRNA BC133493.1 - 8 - 9 1221 71

View Full Product Details