Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UNC93A
Entrez Gene ID
613535
Gene Name
unc-93 homolog A
Gene Aliases
Unc-93A
Chromosome Location
Chr.9: 102000997 - 102035630 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082457.1 NP_001075926.1 4 - 5 788 126
GenBank mRNA BC133579.1 - 4 - 5 788 126

View Full Product Details