Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COMMD10
Entrez Gene ID
538706
Gene Name
COMM domain containing 10
Gene Aliases
-
Chromosome Location
Chr.7: 37598770 - 37783067 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083489.1 NP_001076958.1 1 - 2 49 93
GenBank mRNA BC114666.1 - 1 - 2 49 93

View Full Product Details