Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VSTM2A
Entrez Gene ID
782902
Gene Name
V-set and transmembrane domain containing 2A
Gene Aliases
-
Chromosome Location
Chr.4: 581532 - 607261 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083528.1 NP_001076997.1 1 - 2 89 76
GenBank mRNA BC123836.1 - 1 - 2 89 76

View Full Product Details