Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TMEM25
Entrez Gene ID
504393
Gene Name
transmembrane protein 25
Gene Aliases
-
Chromosome Location
Chr.15: 29198363 - 29203082 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083639.1 NP_001077108.1 1 - 2 263 94
GenBank mRNA BC134598.1 - 1 - 2 263 94

View Full Product Details