Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NEU1
Entrez Gene ID
505554
Gene Name
neuraminidase 1
Gene Aliases
-
Chromosome Location
Chr.23: 27494530 - 27498491 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083642.1 NP_001077111.1 5 - 6 1126 88
GenBank mRNA BC134571.1 - 5 - 6 1126 88

View Full Product Details