Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAP2K3
Entrez Gene ID
516039
Gene Name
mitogen-activated protein kinase kinase 3
Gene Aliases
-
Chromosome Location
Chr.19: 35255910 - 35273754 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083693.1 NP_001077162.1 1 - 2 202 105
GenBank mRNA BC134585.1 - 1 - 2 202 105

View Full Product Details