Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM255A
Entrez Gene ID
533569
Gene Name
transmembrane protein 255A
Gene Aliases
FAM70A
Chromosome Location
Chr.X: 4827858 - 4875893 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083721.1 NP_001077190.1 1 - 2 155 65
GenBank mRNA BC134568.1 - 1 - 2 155 65

View Full Product Details