Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LY86
Entrez Gene ID
613856
Gene Name
lymphocyte antigen 86
Gene Aliases
-
Chromosome Location
Chr.23: 48528635 - 48576190 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083761.1 NP_001077230.1 4 - 5 493 81
GenBank mRNA BC134752.1 - 4 - 5 493 81

View Full Product Details