Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C11H2orf68
Entrez Gene ID
616899
Gene Name
chromosome 11 C2orf68 homolog
Gene Aliases
-
Chromosome Location
Chr.11: 49397862 - 49403918 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083775.1 NP_001077244.1 1 - 2 170 57
GenBank mRNA BC134730.1 - 1 - 2 170 57

View Full Product Details