Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C11H2orf68
Entrez Gene ID
616899
Gene Name
chromosome 11 C2orf68 homolog
Gene Aliases
-
Chromosome Location
Chr.11: 49397862 - 49403918 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083775.1 NP_001077244.1 2 - 3 295 70
GenBank mRNA BC134730.1 - 2 - 3 295 70

View Full Product Details