Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BTC
Entrez Gene ID
280737
Gene Name
betacellulin
Gene Aliases
-
Chromosome Location
Chr.6: 89681969 - 89731873 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173896.2 NP_776321.2 1 - 2 114 104
GenBank mRNA BC133541.1 - 1 - 2 114 104

View Full Product Details