Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAPRE2
Entrez Gene ID
510159
Gene Name
microtubule associated protein RP/EB family member 2
Gene Aliases
-
Chromosome Location
Chr.24: 21874959 - 22062348 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034374.2 NP_001029546.1 5 - 6 883 112
GenBank mRNA BC102764.1 - 5 - 6 897 112
DV848567.1 - 5 - 6 701 112
DY163084.1 - 1 - 2 67 112

View Full Product Details