Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SAMD3
Entrez Gene ID
511666
Gene Name
sterile alpha motif domain containing 3
Gene Aliases
-
Chromosome Location
Chr.9: 68288224 - 68334821 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038083.3 NP_001033172.2 6 - 7 762 64
GenBank mRNA BC111302.1 - 6 - 7 763 64

View Full Product Details