Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PIP5KL1
Entrez Gene ID
523736
Gene Name
phosphatidylinositol-4-phosphate 5-kinase like 1
Gene Aliases
-
Chromosome Location
Chr.11: 98588917 - 98598087 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075847.2 NP_001069315.2 3 - 4 342 103
GenBank mRNA BC118210.1 - 3 - 4 342 103

View Full Product Details