Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PIP5KL1
Entrez Gene ID
523736
Gene Name
phosphatidylinositol-4-phosphate 5-kinase like 1
Gene Aliases
-
Chromosome Location
Chr.11: 98588917 - 98598087 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075847.2 NP_001069315.2 5 - 6 621 57
GenBank mRNA BC118210.1 - 5 - 6 621 57

View Full Product Details