Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRMT61A
Entrez Gene ID
510283
Gene Name
tRNA methyltransferase 61A
Gene Aliases
GCD14, TRM61
Chromosome Location
Chr.21: 68166399 - 68172831 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098927.2 NP_001092397.1 2 - 3 416 95
GenBank mRNA BC146157.1 - 2 - 3 418 95
BC151461.1 - 2 - 3 402 95

View Full Product Details