Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
L2HGDH
Entrez Gene ID
514230
Gene Name
L-2-hydroxyglutarate dehydrogenase
Gene Aliases
-
Chromosome Location
Chr.10: 43166823 - 43219854 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101090.1 NP_001094560.1 3 - 4 467 101
GenBank mRNA BC151577.1 - 3 - 4 467 101

View Full Product Details