Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NINJ2
Entrez Gene ID
509556
Gene Name
ninjurin 2
Gene Aliases
-
Chromosome Location
Chr.5: 107436493 - 107509080 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101914.2 NP_001095384.1 1 - 2 318 93
GenBank mRNA BC148125.1 - 1 - 2 319 93

View Full Product Details