Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KANK4
Entrez Gene ID
519027
Gene Name
KN motif and ankyrin repeat domains 4
Gene Aliases
-
Chromosome Location
Chr.3: 83258461 - 83334028 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102053.2 NP_001095523.2 5 - 6 2597 77
GenBank mRNA BC149084.1 - 7 - 8 1110 77

View Full Product Details