Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADH4
Entrez Gene ID
520508
Gene Name
alcohol dehydrogenase 4 (class II), pi polypeptide
Gene Aliases
-
Chromosome Location
Chr.6: 25480627 - 25512888 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102059.2 NP_001095529.1 8 - 9 1207 112
GenBank mRNA BC149290.1 - 8 - 9 1208 112

View Full Product Details