Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STARD7
Entrez Gene ID
524966
Gene Name
StAR related lipid transfer domain containing 7
Gene Aliases
-
Chromosome Location
Chr.11: 2318507 - 2338037 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102086.2 NP_001095556.1 6 - 7 662 70
GenBank mRNA BC148872.1 - 6 - 7 663 70

View Full Product Details