Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRDM8
Entrez Gene ID
507890
Gene Name
PR/SET domain 8
Gene Aliases
-
Chromosome Location
Chr.6: 94868837 - 94888990 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103232.1 NP_001096702.1 2 - 3 465 69
GenBank mRNA BC151684.1 - 2 - 3 465 69
BC151777.1 - 2 - 3 466 69

View Full Product Details