Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EFHD2
Entrez Gene ID
514259
Gene Name
EF-hand domain family member D2
Gene Aliases
-
Chromosome Location
Chr.16: 52057201 - 52075315 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103245.1 NP_001096715.1 2 - 3 511 63
GenBank mRNA BC140480.1 - 2 - 3 511 63

View Full Product Details