Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LORICRIN
Entrez Gene ID
505154
Gene Name
loricrin cornified envelope precursor protein
Gene Aliases
LOR
Chromosome Location
Chr.3: 17212104 - 17214637 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113757.1 NP_001107229.1 1 - 2 35 75
GenBank mRNA BC118285.1 - 1 - 2 35 75

View Full Product Details