Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HAMP
Entrez Gene ID
512301
Gene Name
hepcidin antimicrobial peptide
Gene Aliases
hepcidin
Chromosome Location
Chr.18: 45994911 - 45996358 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114508.2 NP_001107980.1 2 - 3 201 75
GenBank mRNA AB445457.1 - 2 - 3 130 75
BC111658.1 - 2 - 3 202 75

View Full Product Details