The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098136.1 | NP_001091605.1 | 8 - 9 | 1309 | 58 |
GenBank mRNA | BC140491.1 | - | 8 - 9 | 1309 | 58 |