Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GLRA4
Entrez Gene ID
531678
Gene Name
glycine receptor alpha 4 (pseudogene)
Gene Aliases
-
Chromosome Location
Chr.X: 52452361 - 52478436 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099049.1 NP_001092519.1 4 - 5 862 61
GenBank mRNA BC142201.1 - 4 - 5 862 61

View Full Product Details