Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LYG2
Entrez Gene ID
536852
Gene Name
lysozyme g2
Gene Aliases
-
Chromosome Location
Chr.11: 4366149 - 4377072 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099076.1 NP_001092546.1 3 - 4 454 72
GenBank mRNA BC142237.1 - 3 - 4 454 72

View Full Product Details