Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KRT26
Entrez Gene ID
539216
Gene Name
keratin 26
Gene Aliases
CK-26, K26
Chromosome Location
Chr.19: 40963731 - 40969876 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099096.1 NP_001092566.1 6 - 7 1210 114
GenBank mRNA BC146074.1 - 6 - 7 1210 114

View Full Product Details