Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLK6
Entrez Gene ID
532677
Gene Name
kallikrein related peptidase 6
Gene Aliases
KLNI
Chromosome Location
Chr.18: 56945609 - 56954276 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101140.2 NP_001094610.1 5 - 6 671 96
GenBank mRNA BC149713.1 - 5 - 6 672 96

View Full Product Details