Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VSTM1
Entrez Gene ID
618514
Gene Name
V-set and transmembrane domain containing 1
Gene Aliases
-
Chromosome Location
Chr.18: 61686401 - 61717843 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101274.2 NP_001094744.1 1 - 2 227 91
GenBank mRNA BC149691.1 - 1 - 2 228 91

View Full Product Details