Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SALL2
Entrez Gene ID
527574
Gene Name
spalt like transcription factor 2
Gene Aliases
-
Chromosome Location
Chr.10: 25641299 - 25655398 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC149892.1 - 2 - 3 503 69

View Full Product Details