Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BOLL
Entrez Gene ID
527841
Gene Name
boule homolog, RNA binding protein
Gene Aliases
-
Chromosome Location
Chr.2: 86226318 - 86295179 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102115.1 NP_001095585.1 4 - 5 452 116
NM_001168704.1 NP_001162175.1 4 - 5 416 116
GenBank mRNA BC149292.1 - 4 - 5 452 116
EU050657.1 - 4 - 5 364 116
FJ623266.1 - 4 - 5 367 116
GU187434.1 - 4 - 5 416 116
GU187435.1 - 4 - 5 452 116

View Full Product Details