Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IYD
Entrez Gene ID
534110
Gene Name
iodotyrosine deiodinase
Gene Aliases
-
Chromosome Location
Chr.9: 87263945 - 87509745 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102165.1 NP_001095635.1 2 - 3 423 86
GenBank mRNA BC151488.1 - 2 - 3 423 86

View Full Product Details