Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NAGLU
Entrez Gene ID
789125
Gene Name
N-acetyl-alpha-glucosaminidase
Gene Aliases
-
Chromosome Location
Chr.19: 42618464 - 42625472 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102226.2 NP_001095696.1 2 - 3 565 120
GenBank mRNA BC148147.1 - 2 - 3 566 120

View Full Product Details