Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NOP2
Entrez Gene ID
541169
Gene Name
NOP2 nucleolar protein
Gene Aliases
NOL1
Chromosome Location
Chr.5: 103848151 - 103856941 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102257.1 NP_001095727.1 5 - 6 574 72
GenBank mRNA BC149978.1 - 5 - 6 574 72
BC151441.1 - 5 - 6 574 72

View Full Product Details